Learn More
Medchemexpress LLC 9-acridinecarboxylic acid | 5336-90-3 | MFCD00009734 | 99.9% | 223.23 g/mol | C14H9NO2 | 10 G
SDP
Catalog No. 5000301581
Encompass_Preferred
9-Acridinecarboxylic acid is an acridine-derived carboxylic acid used as a chemical intermediate and precursor in the synthesis of cholinesterase inhibitors. It is used in medicinal chemistry research to prepare derivatives reported to have nanomolar inhibitory potency against acetylcholinesterase (AChE) and butyrylcholinesterase (BChE). Typical supplied purity is approximately 99.9%, and the compound is available in multiple laboratory pack sizes.
  • Precursor for synthesis of cholinesterase inhibitors.
  • Derivatives reported to show nanomolar potency against AChE and BChE.
  • High purity (≈99.9%) suitable for research applications.
  • Available in multiple small-scale pack sizes for laboratory use.
  • Molecular formula C14H9NO2 and molecular weight 223.23 g/mol.
Catalog No. 50-003-01581 Supplier Medchemexpress LLC Supplier No. HYW02275910G
9-Acridinecarboxylic acid is an acridine-derived carboxylic acid used as a chemical intermediate and precursor in the synthesis of cholinesterase inhibitors. It is used in medicinal chemistry research to prepare derivatives reported to have nanomolar inhibitory potency against acetylcholinesterase (AChE) and butyrylcholinesterase (BChE). Typical supplied purity is approximately 99.9%, and the compound is available in multiple laboratory pack sizes.
  • Precursor for synthesis of cholinesterase inhibitors.
  • Derivatives reported to show nanomolar potency against AChE and BChE.
  • High purity (≈99.9%) suitable for research applications.
  • Available in multiple small-scale pack sizes for laboratory use.
  • Molecular formula C14H9NO2 and molecular weight 223.23 g/mol.