Learn More
Medchemexpress LLC 9-acridinecarboxylic acid | 5336-90-3 | MFCD00009734 | 99.9% | 223.23 g/mol | C14H9NO2 | 25 G
SDP
Catalog No. 5000303982
Encompass_Preferred
9-Acridinecarboxylic acid is an acridine-derived carboxylic acid used as a research reagent and synthetic intermediate. It is reported as a precursor for cholinesterase inhibitors and is used in medicinal chemistry studies investigating AChE and BChE inhibition.
  • Precursor for synthesis of cholinesterase inhibitors.
  • Reported high chemical purity suitable for research applications.
  • Molecular formula C14H9NO2; molecular weight 223.23 g/mol.
  • Available in multiple lab-scale package sizes, including 25 g.
  • Suitable as an intermediate in medicinal chemistry and inhibitor development.
Catalog No. 50-003-03982 Supplier Medchemexpress LLC Supplier No. 25GHYW022759
Add to Cart
Add to Cart
9-Acridinecarboxylic acid is an acridine-derived carboxylic acid used as a research reagent and synthetic intermediate. It is reported as a precursor for cholinesterase inhibitors and is used in medicinal chemistry studies investigating AChE and BChE inhibition.
  • Precursor for synthesis of cholinesterase inhibitors.
  • Reported high chemical purity suitable for research applications.
  • Molecular formula C14H9NO2; molecular weight 223.23 g/mol.
  • Available in multiple lab-scale package sizes, including 25 g.
  • Suitable as an intermediate in medicinal chemistry and inhibitor development.