Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate

Catalog No. A799225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A799225G 5 g
A799221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A799225G Supplier Sigma Aldrich Supplier No. A799225G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate

Specifications

CAS 206658-92-6
Melting Point 284°C to 286°C (lit.)
Molecular Formula C13H14N2 -+ HCl -+ xH2O
Linear Formula C13H14N2 -+ HCl -+ xH2O
MDL Number MFCD00150067
Quantity 5 g
Synonym THA hydrochloride hydrate; Tacrine hydrochloride; Tetrahydroaminacrine hydrochloride hydrate
Molecular Weight (g/mol) 234.72 (anhydrous basis)
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.