Learn More
Medchemexpress LLC 9-amino-6-chloro-2-methoxyacridine | 602166 | 99.6% | 258.70 | C14H11ClN2O | 10 MG
SDP
Catalog No. 5000228686
Encompass_Preferred
9-Amino-6-chloro-2-methoxyacridine (ACMA) is a pH-sensitive fluorescent probe used to monitor changes in vacuolar and vesicular pH and to follow proton translocation across membranes in biochemical assays.
  • pH-sensitive fluorescent probe for monitoring vacuolar and vesicular pH.
  • Useful in proton translocation and H+/solute antiport assays.
  • High purity (~99.6%) suitable for sensitive fluorescence measurements.
  • Available in small reagent quantities (e.g., 10 mg) for laboratory assays.
  • Supplied with supporting documentation such as SDS, COA, HNMR, and LCMS.
Catalog No. 50-002-28686 Supplier Medchemexpress LLC Supplier No. HY11815510MG
9-Amino-6-chloro-2-methoxyacridine (ACMA) is a pH-sensitive fluorescent probe used to monitor changes in vacuolar and vesicular pH and to follow proton translocation across membranes in biochemical assays.
  • pH-sensitive fluorescent probe for monitoring vacuolar and vesicular pH.
  • Useful in proton translocation and H+/solute antiport assays.
  • High purity (~99.6%) suitable for sensitive fluorescence measurements.
  • Available in small reagent quantities (e.g., 10 mg) for laboratory assays.
  • Supplied with supporting documentation such as SDS, COA, HNMR, and LCMS.