Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-amino-6-chloro-2-methoxyacridine | 602166 | MFCD07374304 | 99.6% | 258.70 | C14H11ClN2O | 5 MG
SDP

Catalog No. 5000226913
Encompass_Preferred

9-Amino-6-chloro-2-methoxyacridine (ACMA) is a pH-sensitive fluorescent probe used to monitor changes in vacuolar and membrane compartment pH and to study proton gradients and H+/solute antiport activity in biological assays.

  • pH-sensitive fluorescent probe suitable for monitoring vacuolar and membrane pH.
  • High purity (99.56%), suitable for analytical and research applications.
  • Molecular weight 258.70 and formula C14H11ClN2O.
  • Offered as small-mass solids (5 mg, 10 mg, 25 mg) and as 10 mM solutions in DMSO.
  • Commonly used in assays of proton gradients and H+/solute antiport systems.

Catalog No. 50-002-26913 Supplier Medchemexpress LLC Supplier No. HY1181555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

9-Amino-6-chloro-2-methoxyacridine (ACMA) is a pH-sensitive fluorescent probe used to monitor changes in vacuolar and membrane compartment pH and to study proton gradients and H+/solute antiport activity in biological assays.

  • pH-sensitive fluorescent probe suitable for monitoring vacuolar and membrane pH.
  • High purity (99.56%), suitable for analytical and research applications.
  • Molecular weight 258.70 and formula C14H11ClN2O.
  • Offered as small-mass solids (5 mg, 10 mg, 25 mg) and as 10 mM solutions in DMSO.
  • Commonly used in assays of proton gradients and H+/solute antiport systems.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.