Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-amino-6-chloro-2-methoxyacridine | 602166 | MFCD07374304 | 99.6% | 258.70 g/mol | C14H11ClN2O | 1 ML
SDP

Catalog No. 5000307716
Encompass_Preferred

9-Amino-6-chloro-2-methoxyacridine is a pH-sensitive fluorescent probe used to monitor intravesicular and vacuolar pH changes. It is commonly applied to detect proton gradients and to assess H+/solute antiport or membrane transport activity by reporting fluorescence changes in response to pH shifts.

  • pH-sensitive fluorescent probe for monitoring vacuolar and vesicular pH.
  • Detects proton gradients and H+/solute antiport activity via fluorescence changes.
  • Supplied as a 10 mM solution in DMSO (1 mL) or as solid forms for flexibility.
  • High purity suitable for biochemical assays (manufacturer reports 99.56%).
  • Molecular formula C14H11ClN2O; molecular weight 258.70 g/mol.

Catalog No. 50-003-07716 Supplier Medchemexpress LLC Supplier No. HY11815510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-Amino-6-chloro-2-methoxyacridine is a pH-sensitive fluorescent probe used to monitor intravesicular and vacuolar pH changes. It is commonly applied to detect proton gradients and to assess H+/solute antiport or membrane transport activity by reporting fluorescence changes in response to pH shifts.

  • pH-sensitive fluorescent probe for monitoring vacuolar and vesicular pH.
  • Detects proton gradients and H+/solute antiport activity via fluorescence changes.
  • Supplied as a 10 mM solution in DMSO (1 mL) or as solid forms for flexibility.
  • High purity suitable for biochemical assays (manufacturer reports 99.56%).
  • Molecular formula C14H11ClN2O; molecular weight 258.70 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.