Learn More
TARGETMOL CHEMICALS INC 9-AMINO-CPT 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3S195710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 9-amino-CPT (Aminocamptothecin) is an inhibitor of topoisomerase I with potent anticancer activity. purity: 99%
Catalog No. 50-258-0010