Learn More
Medchemexpress LLC 9-Amino-NeuAc | 153232-60-1 | C11H20N2O8 | 1 MG
SDP
Catalog No. 5000372783
Encompass_Preferred
9-Amino-NeuAc is a substrate of synthase that can be converted to CMP9-amino-NeuAc, which is then activated to the corresponding CMPglycoside and transferred to asialoglycoprotein. This compound can also be used to target liposome synthesis while modulating tumor surface immunogenicity.
  • Functions as a substrate of synthase
  • Converts to CMP9-amino-NeuAc
  • Activated to CMPglycoside
  • Transferred to asialoglycoprotein
  • Targets liposome synthesis
  • Modulates tumor surface immunogenicity
Catalog No. 50-003-72783 Supplier Medchemexpress LLC Supplier No. HY1535451MG
9-Amino-NeuAc is a substrate of synthase that can be converted to CMP9-amino-NeuAc, which is then activated to the corresponding CMPglycoside and transferred to asialoglycoprotein. This compound can also be used to target liposome synthesis while modulating tumor surface immunogenicity.
  • Functions as a substrate of synthase
  • Converts to CMP9-amino-NeuAc
  • Activated to CMPglycoside
  • Transferred to asialoglycoprotein
  • Targets liposome synthesis
  • Modulates tumor surface immunogenicity