Learn More
Apexbio Technology LLC 9-AMINOACRIDINE-200MG
SDP
Catalog No. 5000568762
Encompass_Preferred
9-Aminoacridine (CAS No 90-45-9) is a polycyclic compound primarily used in biomedical research particularly in molecular biology and pharmacology Originating as a synthetic derivative of acridine it exhibits its mechanism of action through intercalation with DNA disrupting cellular processes such as replication and transcription This compound is effective in a nanomolar to low micromolar range in vitro indicating its potent bioactivity Common applications include studies on bacterial and eukaryotic cell models and it is also utilized in drug screening assays Researchers often use it in experimental concentrations that depend on specific research designs allowing for flexibility in various experimental contexts Additionally 9-Aminoacridine s fluorescent properties make it valuable in studying nucleic acid interactions and tracking cellular events in both in vitro and in vivo settings
Catalog No. 50-005-68762 Supplier Apexbio Technology LLC Supplier No. C6567200
9-Aminoacridine (CAS No 90-45-9) is a polycyclic compound primarily used in biomedical research particularly in molecular biology and pharmacology Originating as a synthetic derivative of acridine it exhibits its mechanism of action through intercalation with DNA disrupting cellular processes such as replication and transcription This compound is effective in a nanomolar to low micromolar range in vitro indicating its potent bioactivity Common applications include studies on bacterial and eukaryotic cell models and it is also utilized in drug screening assays Researchers often use it in experimental concentrations that depend on specific research designs allowing for flexibility in various experimental contexts Additionally 9-Aminoacridine s fluorescent properties make it valuable in studying nucleic acid interactions and tracking cellular events in both in vitro and in vivo settings