Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Aminoacridine hemihydrate, 98%, Thermo Scientific™

Catalog No. AC134410250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC134410250 25 g
AC134410050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC134410250 Supplier Thermo Scientific Chemicals Supplier No. 134410250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65944-23-2
Molecular Formula C26H22N4O
Molecular Weight (g/mol) 406.489
InChI Key SPWFQHCZHQCDAG-UHFFFAOYSA-N
Synonym acridin-9-amine hydrate, 9-aminoacridine hemihydrate
PubChem CID 2723763
IUPAC Name acridin-9-amine;hydrate
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N.C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N.O

Specifications

Melting Point 230.0°C to 235.0°C
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, freely soluble in alcohol, slightly soluble in chloroform, toluene and, pyridine
Formula Weight 203.16
Chemical Name or Material 9-Aminoacridine hemihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.