Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Aminoacridine, Matrix Substance for MALDI-MS, ≥99.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015863 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5863 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5863 Supplier MilliporeSigma Supelco Supplier No. 928171G
Add to Cart
Add to Cart

9-Aminoacridine, an aminoacridine dye which acts as a potent mutagen in viruses and bacteria.

TRUSTED_SUSTAINABILITY

Specifications

CAS 90-45-9
Linear Formula C13H10N2
MDL Number MFCD00037839
Melting Point 238°C to 240°C
Synonym Aminacrine
Type Matrix Substance for MALDI-MS
Quantity 1 g
Formula Weight 194.23
Percent Purity ≥99.5% (HPLC)
Grade Matrix Substance for MALDI-MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.