Learn More
Medchemexpress LLC 9-Aminononanoic acid 250mg
SDP
Catalog No. 5000205996
Encompass_Preferred
9-Aminononanoic acid can be used as a PROTAC linker and other conjugation in the synthesis of PROTACs 9-Aminononanoic acid is an alkane chain with terminal carboxlic acid and amine groups The amino group (NH2) is reactive with carboxylic acids activated NHS esters carbonyls (ketone aldehyde) etc The terminal carboxylic acid can react with primary amine groups of activated NHS ester to form a stable amide bond
Catalog No. 50-002-05996 Supplier Medchemexpress LLC Supplier No. 250MGHYW105727
9-Aminononanoic acid can be used as a PROTAC linker and other conjugation in the synthesis of PROTACs 9-Aminononanoic acid is an alkane chain with terminal carboxlic acid and amine groups The amino group (NH2) is reactive with carboxylic acids activated NHS esters carbonyls (ketone aldehyde) etc The terminal carboxylic acid can react with primary amine groups of activated NHS ester to form a stable amide bond