Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Anthracenecarboxaldehyde

Numéro de catalogue. 27868825G missing translation for 'brandsLinkText'
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
27-868-825G 25 g
27-868-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 27-868-825G Fournisseur Sigma Aldrich Code fournisseur 27868825G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

9-Anthracenecarboxaldehyde

Spécifications

CAS 642-31-9
Melting Point 103°C to 105°C (lit.)
Molecular Formula C15H10O
Linear Formula C15H10O
MDL Number MFCD00001254
Quantity 25 g
Synonym 9-Anthraldehyde
Molecular Weight (g/mol) 206.24
Percent Purity 97%
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.