Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Anthracenemethanol

Catalog No. 18724050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-724-050G 50 g
18-724-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-724-050G Supplier Sigma Aldrich Supplier No. 18724050G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 9-anthracenemethanol participates in ring-opening polymerization of L-lactide catalyzed by alumoxane. it undergoes proton exchange reaction with potassium tert-butoxide to yield potassium 9-anthracenemethoxide.9-anthracenemethanol can be used:
  • As a starting material to prepare 9-anthracenylmethyl-1-piperazinecarboxylate, which acts as a reagent in the determination of isocyanates using HPLC
  • In the diels-alder reaction with dimethylacetylene-dicarboxylate to yield lactone derivatives
  • As an initiator in the ring-opening polymerization of α-valerolactone to yield poly(α-valerolactone)
  • As a starting material in the synthesis of polymer-supported anthracene, which acts as a dienophile scavenger in cycloaddition reactions

Specifications

CAS 1468-95-7
Molecular Formula C15H12O
Linear Formula C15H12O
MDL Number MFCD00001264
Quantity 50 g
Synonym 9-(Hydroxymethyl)anthracene
Molecular Weight (g/mol) 208.26
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.