Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Benzhydrylidene-10-anthrone, Thermo Scientific™

Catalog No. AC105400010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC105400010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC105400010 Supplier Thermo Scientific Chemicals Supplier No. 105400010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 667-91-4
Molecular Formula C27H18O
Molecular Weight (g/mol) 358.44
MDL Number MFCD00019172
InChI Key SPOJSTUMWGMCFP-UHFFFAOYSA-N
Synonym 9-benzhydrylidene-10-anthrone, 10-diphenylmethylene anthracen-9 10h-one, 10-diphenylmethylidene anthracen-9-one, anthrafuchson, 10-benzhydrylidene-9-anthrone, 10-diphenylmethylene anthracen-9-one, 9-diphenylmethylene-10 9h-anthracenone, 9 10h-anthracenone,10-diphenylmethylene, 10-diphenylmethylene-9 10h-anthracenone #, 9 10h-anthracenone, 10-diphenylmethylene
PubChem CID 69586
IUPAC Name 10-benzhydrylideneanthracen-9-one
SMILES C1=CC=C(C=C1)C(=C2C3=CC=CC=C3C(=O)C4=CC=CC=C42)C5=CC=CC=C5

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 200°C to 204°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 1 g
Beilstein 07, 551
Formula Weight 358.44
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzhydrylidene-10-anthrone
Show More Show Less
EINECSNumber 211-570-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.