Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Benzyl-2-bromo-9H-carbazole 98.0+%, TCI America™
SDP

Catalog No. B54605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54605G 5 g
B54601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54605G Supplier TCI America Supplier No. B54605G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1401863-51-1
Molecular Formula C19H14BrN
Molecular Weight (g/mol) 336.232
InChI Key BFHNUHBRWXASFQ-UHFFFAOYSA-N
PubChem CID 123133636
IUPAC Name 9-benzyl-2-bromocarbazole
SMILES C1=CC=C(C=C1)CN2C3=CC=CC=C3C4=C2C=C(C=C4)Br

Specifications

Melting Point 135°C
Color White
Quantity 5 g
Formula Weight 336.23
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzyl-2-bromo-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.