Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Benzylcarbazole-3,6-dicarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. B2805100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2805100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2805100MG Supplier TCI America Supplier No. B2805100MG

Chemical Identifiers

CAS 200698-05-1
Molecular Formula C21H15NO2
Molecular Weight (g/mol) 313.356
InChI Key RZSXZJXJVWYRIU-UHFFFAOYSA-N
Synonym 9-Benzyl-3,6-diformylcarbazole
PubChem CID 18931891
IUPAC Name 9-benzylcarbazole-3,6-dicarbaldehyde
SMILES C1=CC=C(C=C1)CN2C3=C(C=C(C=C3)C=O)C4=C2C=CC(=C4)C=O

Specifications

Quantity 100 mg
Formula Weight 313.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzylcarbazole-3,6-dicarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.