Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-beta-D-Arabinofuranosyl-2-fluoroadenine-5'-monophosphate, 99%, Thermo Scientific™

Catalog No. AAJ65154MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65154MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65154MD Supplier Thermo Scientific Chemicals Supplier No. J65154MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in DMSO or water at 5mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75607-67-9
Molecular Formula C10H13FN5O7P
Molecular Weight (g/mol) 365.21
MDL Number MFCD00866418
InChI Key GIUYCYHIANZCFB-FJFJXFQQSA-N
Synonym fludarabine phosphate, fludara, fludarabine 5'-monophosphate, fludura, oforta, 2-fluoro-ara amp, fludarabine monophosphate, famp, unii-1x9vk9o1sc
PubChem CID 30751
ChEBI CHEBI:63599
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC(F)=N1

Specifications

Color White
Quantity 250 mg
Solubility Information Soluble in DMSO or water at 5mg/ml
IUPAC Name {[(2R,3S,4S,5R)-5-(6-amino-2-fluoro-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Formula Weight 365.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 9-beta-D-Arabinofuranosyl-2-fluoroadenine-5'-monophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UO7440900
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.