Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-beta-D-Arabinofuranosyladenine-5'-monophosphate, 99%, Thermo Scientific Chemicals

Catalog No. AAJ6565306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6565306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6565306 Supplier Thermo Scientific Chemicals Supplier No. J6565306

missing translation for 'validMainframeMsg'

CAS: 29984-33-6 | Molecular Formula: C10H14N5O7P | Molecular Weight: 347.22 g/mol

Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor.

Solubility
Soluble in water at 4mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29984-33-6
Molecular Formula C10H14N5O7P
Molecular Weight (g/mol) 347.22
MDL Number MFCD00069723
InChI Key UDMBCSSLTHHNCD-UHTZMRCNSA-N
Synonym vidarabine monophosphate, ara-amp, vidarabine phosphate, adenine arabinoside monophosphate, 9-5-o-phosphono-beta-d-arabinofuranosyl adenine, arabinofuranosyladenine monophosphate, vidarabine 5'-monophosphate, vidarabine-5'-monophosphate, vidarabine phosphate usan, arabinosyladenine monophosphate
PubChem CID 34768
IUPAC Name [(2R,3S,4S,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC=N1

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in water at 4mg/ml
Formula Weight 347.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 9-beta-D-Arabinofuranosyladenine-5'-monophosphate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 249-990-8
RTECSNumber AU6204000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.