Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Borabicyclo[3.3.1]Nonane Solution

Catalog No. 459496100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-949-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 45-949-6100ML Supplier Sigma Aldrich Supplier No. 459496100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protecting group foralkenes†Reactant for: Linear SPPS synthesis of ubiquitin derivativesCopper-catalyzed cross-coupling reactions of organoboron compounds with primary alkyl halides and pseudohalidesIntramolecular insertion of alkenes into palladium-nitrogen bondsPreparation of (phosphonoacetyl)ornithine to study effect on arginine biosynthetic genes in yeastHetero-Diels-Alder reaction for synthesis of spirocyclic alkaloids

Specifications

CAS 5565-32-2
Melting Point 50°C to 52°C
Molecular Formula C9H27ClSi4
Linear Formula [(CH3)3Si]3SiCl
MDL Number MFCD01321222
Quantity 100 mL
Synonym 9-BBN
Molecular Weight (g/mol) 283.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.