Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Borabicyclo[3.3.1]Nonane Solution

Catalog No. 151076800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
8 L
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-107-6800ML 800 mL
15-107-68L 8 L
15-107-618LKL 18 L
50-308-1430 4 x 25 mL
15-107-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 15-107-6800ML Supplier Sigma Aldrich Supplier No. 151076800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

9-Borabicyclo[3.3.1]nonane is an organic compound, which is generally used as a hydroborating agent. It is thermally stable, less sensitive to oxygen and water when compared to other dialkyl boranes. It is commonly used to prepare aldehydes from alkynes.Selective hydroboration reduction reagent.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 280-64-8
Density 0.894 g/mL at 25°C
Molecular Formula C8H15B
Linear Formula C8H15B
MDL Number MFCD00074742
Quantity 800 mL
Synonym 9-BBN
Molecular Weight (g/mol) 122.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.