Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromo-10-[4-(1-naphthyl)phenyl]anthracene 98.0+%, TCI America™
SDP

Catalog No. B45701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45701G 1 g
B4570200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45701G Supplier TCI America Supplier No. B45701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1092390-01-6
Molecular Formula C30H19Br
Molecular Weight (g/mol) 459.386
MDL Number MFCD20486477
InChI Key AEIPFESDNDUXNK-UHFFFAOYSA-N
PubChem CID 59861528
IUPAC Name 9-bromo-10-(4-naphthalen-1-ylphenyl)anthracene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=CC=C(C=C3)C4=C5C=CC=CC5=C(C6=CC=CC=C64)Br

Specifications

Melting Point 213°C
Quantity 1 g
Formula Weight 459.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-[4-(1-naphthyl)phenyl]anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.