Learn More
9-Bromo-10-phenylanthracene, 98%
Description
It is used as an OLED material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as an OLED material.
Solubility
Insoluble in water. Soluble in hot toluene, dimethylformamide.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 154°C to 158°C |
| Color | Yellow |
| Quantity | 250 mg |
| Beilstein | 2277064 |
| Solubility Information | Insoluble in water. Soluble in hot toluene,dimethylformamide. |
| Formula Weight | 333.23 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | 9-Bromo-10-phenylanthracene |
Safety and Handling
| Hazard Category | H302-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H317-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.