Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Bromoanthracene

Catalog No. B566095G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B566095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B566095G Supplier Sigma Aldrich Supplier No. B566095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant involved in:
  • Synthesis of meta- and paracyclophanes containing unsaturated amino acids
  • Intramolecular friedel-crafts reactions for synthesis of 1-tetralonesinvolved in investigations of pharmacological activity pertaining to:
  • Apoptosis, proliferation, and histon deacetylase activity in human colorectal cancer cells
  • Portable albumin binders for extending the half-life of contrast agents

Specifications

CAS 1564-64-3
Melting Point 97°C to 100°C (lit.)
Molecular Formula C14H9Br
Linear Formula C14H9Br
MDL Number MFCD00001243
Quantity 5 g
Molecular Weight (g/mol) 257.13
Percent Purity 94%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.