Learn More
Medchemexpress LLC 9-chloro-2-ethyl-1-methyl-3-(2,2,2-trifluoroethyl)-3H,6H,7H-pyrrolo[3,2-f]quinolin-7-one | 917891-35-1 | ≥95.0% | 342.74 g/mol | C16H14ClF3N2O | 50 MG
SDP
Catalog No. 5000245017
Encompass_Preferred
LGD-3303 is a nonsteroidal selective androgen receptor modulator (SARM) provided for laboratory research use. It is used in preclinical studies to investigate androgen receptor signaling, and to study muscle and bone biology. Research use only; not for human or veterinary applications.
  • Selective androgen receptor modulator for preclinical AR studies.
  • Applicable to muscle and bone biology research.
  • Available in small research pack sizes for assay development.
  • Stable at -20 °C for long-term storage.
  • Typical purity ≥95.0% supports biochemical assays.
Catalog No. 50-002-45017 Supplier Medchemexpress LLC Supplier No. HY10357650MG
LGD-3303 is a nonsteroidal selective androgen receptor modulator (SARM) provided for laboratory research use. It is used in preclinical studies to investigate androgen receptor signaling, and to study muscle and bone biology. Research use only; not for human or veterinary applications.
  • Selective androgen receptor modulator for preclinical AR studies.
  • Applicable to muscle and bone biology research.
  • Available in small research pack sizes for assay development.
  • Stable at -20 °C for long-term storage.
  • Typical purity ≥95.0% supports biochemical assays.