Learn More
Medchemexpress LLC 9-chloro-2-ethyl-1-methyl-3-(2,2,2-trifluoroethyl)-3H-pyrrolo-[3,2-f]quinolin-7(6H)-one | 917891-35-1 | 99.6% | 342.74 g/mol | C16H14ClF3N2O | 10 MG
SDP
Catalog No. 5000247922
Encompass_Preferred
LGD-3303 is a nonsteroidal selective androgen receptor modulator (SARM) used in research to evaluate androgen receptor activation and tissue-selective anabolic effects. It has been characterized in vitro and in vivo for potency at the androgen receptor and for muscle-sparing activity with reduced effects on prostate and other androgen-responsive tissues.
  • Selective androgen receptor modulator (SARM) with potent AR agonist activity.
  • Nonsteroidal compound with reported EC50 ~3.6 nM in transcription assays.
  • Tissue-selective anabolic effects demonstrated in animal studies.
  • High reported purity (99.6%).
  • Supplied with datasheet, safety data sheet, and certificate of analysis.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-47922 Supplier Medchemexpress LLC Supplier No. HY10357610MG
LGD-3303 is a nonsteroidal selective androgen receptor modulator (SARM) used in research to evaluate androgen receptor activation and tissue-selective anabolic effects. It has been characterized in vitro and in vivo for potency at the androgen receptor and for muscle-sparing activity with reduced effects on prostate and other androgen-responsive tissues.
  • Selective androgen receptor modulator (SARM) with potent AR agonist activity.
  • Nonsteroidal compound with reported EC50 ~3.6 nM in transcription assays.
  • Tissue-selective anabolic effects demonstrated in animal studies.
  • High reported purity (99.6%).
  • Supplied with datasheet, safety data sheet, and certificate of analysis.
  • For research use only; not for human or clinical use.