Learn More
Medchemexpress LLC 9-cis-β-Carotene | 13312-52-2 | 99.0% | 536.87 | 500 UG
SDP
Catalog No. 5000351420
Encompass_Preferred
9-cis-β-Carotene is a precursor of retinal, cleaved by beta-carotene oxygenase 1 (BCMO1) to produce 9-cis-retinal. It inhibits photoreceptor degeneration and restores retinal function in vivo, with potential for the study of congenital stationary night blindness and fundus albipunctatus.
  • Inhibits photoreceptor degeneration.
  • Restores retinal function in vivo.
  • Potential for studying congenital stationary night blindness.
  • Potential for studying fundus albipunctatus.
  • Prevents S- and M-cone degeneration in eye cups.
  • Rescues retinal function and preserves cone structure in a mouse model of retinoid cycle defect.
Catalog No. 50-003-51420 Supplier Medchemexpress LLC Supplier No. HY136234500UG
9-cis-β-Carotene is a precursor of retinal, cleaved by beta-carotene oxygenase 1 (BCMO1) to produce 9-cis-retinal. It inhibits photoreceptor degeneration and restores retinal function in vivo, with potential for the study of congenital stationary night blindness and fundus albipunctatus.
  • Inhibits photoreceptor degeneration.
  • Restores retinal function in vivo.
  • Potential for studying congenital stationary night blindness.
  • Potential for studying fundus albipunctatus.
  • Prevents S- and M-cone degeneration in eye cups.
  • Rescues retinal function and preserves cone structure in a mouse model of retinoid cycle defect.