Learn More
Cayman Chemical 9-cIs RetInal 5mg
SDP
Catalog No. 502987637
Encompass_Preferred
A natural retinoid that is produced by oxidation of 9-cis retinol by cRDH; binds to CRBP-I, CRBP-II, and CRALBP (Kds = 8, 5, and 53.3 nM, respectively); inhibits differentiation of RCJ C5.18 chondrogenic cells into cartilage in vitro (IC50 = 8 nM); it rescues cone ERG responses in Irbp-/- mice (0.375 mg)
Catalog No. 50-298-7637 Supplier Cayman Chemical Supplier No. 216925 MG
A natural retinoid that is produced by oxidation of 9-cis retinol by cRDH; binds to CRBP-I, CRBP-II, and CRALBP (Kds = 8, 5, and 53.3 nM, respectively); inhibits differentiation of RCJ C5.18 chondrogenic cells into cartilage in vitro (IC50 = 8 nM); it rescues cone ERG responses in Irbp-/- mice (0.375 mg)