Learn More
Eurofins Discoverx 9-cis-Retinoic acid, 1 mg. (92-1136)
Supplier:  Eurofins Discoverx 921136
Encompass
9-cis-Retinoic acid is a natural metabolite of vitamin A (retinol) and potent agonist of the retinoic acid and retinoid X receptors (Ki=0.5-27 nM and 3.8-12 nM, respectively). Mediates retinol functions for growth and development, and is active in embryonic development by intercellular signaling for the posterior portion of the embryo, and developmental patterning. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9668