Learn More
Cayman Chemical 9-cis Retinoic Acid, Purity: ≥90%, CAS: 5300-03-8, 10 mg
SDP
Catalog No. 502969528
Encompass_Preferred
Potently activates all isoforms of RAR (Ki = 0.5-27 nM) as well as RXR isoforms (Ki = 3.8-12 nM)
Catalog No. 50-296-9528 Supplier Cayman Chemical Supplier No. 1458710 MG
Potently activates all isoforms of RAR (Ki = 0.5-27 nM) as well as RXR isoforms (Ki = 3.8-12 nM)