Learn More
eMolecules​ 5300-03-8 | 9-cis-Retinoic Acid | AA Blocks LLC | MFCD00270072 | 300.442 | C20H28O2 | CC(C=CC1=C(C)CCCC1(C)C)=CC=CC(C)=CC(O)=O | 1mg | 586011463
Catalog No. NC2013720 Shop All eMolecules Inc. Products
Encompass_Preferred
9-cis-Retinoic Acid | AA Blocks LLC | 5300-03-8 | MFCD00270072 | 300.442 | C20H28O2 | CC(C=CC1=C(C)CCCC1(C)C)=CC=CC(C)=CC(O)=O | 1mg | 586011463
Catalog No. NC2013720 Supplier eMolecules​ Supplier No. 586011463
9-cis-Retinoic Acid | AA Blocks LLC | 5300-03-8 | MFCD00270072 | 300.442 | C20H28O2 | CC(C=CC1=C(C)CCCC1(C)C)=CC=CC(C)=CC(O)=O | 1mg | 586011463