Learn More
BIOGEMS INTERNATIONAL INC 9-cis-Retinoic acid 1mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 53003801MG
Encompass
9-cis-Retinoic acid is a potent activator of all isoforms of retinoic acid receptor and the retinoid X receptor, which act as transcription factors that regulate the growth and differentiation of normal and malignant cells.. It is reported to promote oligodendrocyte precursor cell differentiation and myelination and to also inhibit the synthesis of fatty acids and sterols in hepatocytes.
Catalog No. 50-153-0181