Learn More
Enzo Life Sciences 9-cis-Retinoic acid (25mg). CAS: 5300-03-8
SDP
Supplier:  Enzo Life Sciences BMLGR1010025
Encompass_Preferred
A high affinity ligand for the retinoid X receptor (RXR). Destabilizes VDR-RXR heterodimer-DNA complex. Alternative name: Alitretinoin. Purity: ≥98% (UHPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0913