Learn More
Cayman Chemical 9-cis-Retinoic Acid, 5300-03-8, 5 mg
SDP
Supplier:  Cayman Chemical 145875
Encompass_Preferred
Molecular Formula C20H28O2, Purity ≥95%, Formula Weight 300.4, Synonyms: 9-cis-retinoic acid, Alitretinoin, NSC 659772, Panretin, 9-cis RA; Potently activates all isoforms of RAR (Ki = 0.5-27 nM) as well as RXR isoforms (Ki = 3.8-12 nM).
Catalog No. NC0701488