Learn More
Medchemexpress LLC 9-cis-Retinoic acid (Alitretinoin) | 5300-03-8 | C20H28O2 | 100 MG
SDP
Catalog No. 5000375894
Encompass_Preferred
9-cis-Retinoic acid, also known as Alitretinoin, is a vitamin A derivative and a potent RAR/RXR agonist. It induces apoptosis, regulates the cell cycle, and possesses anticancer, anti-inflammatory, and neuroprotection activities.
  • Potent RAR/RXR agonist
  • Induces apoptosis
  • Regulates the cell cycle
  • Exhibits anticancer activity
  • Offers anti-inflammatory effects
  • Provides neuroprotection
Catalog No. 50-003-75894 Supplier Medchemexpress LLC Supplier No. HY15128100MG
9-cis-Retinoic acid, also known as Alitretinoin, is a vitamin A derivative and a potent RAR/RXR agonist. It induces apoptosis, regulates the cell cycle, and possesses anticancer, anti-inflammatory, and neuroprotection activities.
  • Potent RAR/RXR agonist
  • Induces apoptosis
  • Regulates the cell cycle
  • Exhibits anticancer activity
  • Offers anti-inflammatory effects
  • Provides neuroprotection