Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-cis-Retinol acetate | 29584-22-3 | 85.0% | 328.49 | 10 MG
SDP

Catalog No. 5000365348
Encompass_Preferred

9-cis-Retinol acetate is a retinal derivative, also known as 9-cis-Vitamin A acetate or Zuretinol acetate. It is provided as a white to off-white solid and is intended for research use only, not for use with patients.

  • Molecular weight: 328.49
  • Formula: C22H32O2
  • Purity: 85.0%
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-65348 Supplier Medchemexpress LLC Supplier No. 10MGHY16760
May include imposed supplier surcharges.
Add to Cart
Add to Cart

9-cis-Retinol acetate is a retinal derivative, also known as 9-cis-Vitamin A acetate or Zuretinol acetate. It is provided as a white to off-white solid and is intended for research use only, not for use with patients.

  • Molecular weight: 328.49
  • Formula: C22H32O2
  • Purity: 85.0%
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.