Learn More
American Biotech Supply 9 CU. FT. Capacity Premier Flammable Refrigerator and Freezer Combination
Supplier:  American Biotech Supply ABTHCFRFC10
Encompass_Preferred
The Premier Dual Temp Combination Refrigerator / Freezers feature separate microprocessor temperature controller allowing for precise temperature control, verification and recovery. An interior fan driven, forced-draft air circulation system provides excellent temperature uniformity and recovery after door openings.
Catalog No. NC1677037
Request a Quote
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.