Learn More
TARGETMOL CHEMICALS INC 9-CYCLOPENTYLADENINE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T296525MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. 9-Cyclopentyladenine (NSC-19486) is an adenylate cyclase inhibitor that blocks cellular morphological differentiation and phosphorylation of specific signaling molecules. purity: 99%
Catalog No. 50-258-0451