Learn More
Cayman Chemical 9-DeazagunIn 10mg
SDP
Catalog No. 502987612
Encompass_Preferred
An inhibitor of PNP (Kd = 160 nM)
Catalog No. 50-298-7612 Supplier Cayman Chemical Supplier No. 2167410 MG
An inhibitor of PNP (Kd = 160 nM)