Learn More
Cayman Chemical 9-DeazagunIn 50mg
SDP
Catalog No. 502987613
Encompass_Preferred
An inhibitor of PNP (Kd = 160 nM)
Catalog No. 50-298-7613 Supplier Cayman Chemical Supplier No. 2167450 MG
An inhibitor of PNP (Kd = 160 nM)