Learn More
Medchemexpress LLC 9-Decen-1-ol 50g
SDP
Supplier:  Medchemexpress LLC HYW10535950G
Encompass_Preferred
9-Decen-1-ol has a fatty odour This compound is used in the flavor and fragrance industry for its pleasant smell which is often described as sweet floral and fruity It can also be used as an intermediate in the synthesis of various organic compounds including medicine agricultural chemicals and polymer materials Its unique chemical properties make it an important ingredient in many commercial products including soaps detergents and cosmetics
Catalog No. 50-002-06172