Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Decenoic acid, ≥95.0% (GC); 95.0-105.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013114 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3114 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3114 Supplier MilliporeSigma Supelco Supplier No. 44191100MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 14436-32-9
Density 0.918 g/mL (at 25°C)
Flash Point 113°C
Physical Form Neat
Linear Formula H2 C=CH(CH2)7 CO2 H
MDL Number MFCD00036663
Boiling Point 270°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 170.25
Percent Purity ≥95.0% (GC); 95.0 to 105.0% (T)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.