Learn More
Medchemexpress LLC 9-Decyn-1-ol | 17643-36-6 | 1 G
SDP
Catalog No. 5000340581
Encompass_Preferred
9-Decyn-1-ol is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It can be used to conjugate specific compounds to generate potent, highly selective, and PROTAC-based pan-Akt degraders. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables the generation of potent, highly selective, and PROTAC-based pan-Akt degraders
  • Acts as a click chemistry reagent
  • Features an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
Catalog No. 50-003-40581 Supplier Medchemexpress LLC Supplier No. HY1309851G
9-Decyn-1-ol is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It can be used to conjugate specific compounds to generate potent, highly selective, and PROTAC-based pan-Akt degraders. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables the generation of potent, highly selective, and PROTAC-based pan-Akt degraders
  • Acts as a click chemistry reagent
  • Features an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules