Learn More
Medchemexpress LLC 9-Decyn-1-ol | 17643-36-6 | 5 G
SDP
Catalog No. 5000340673
Encompass_Preferred
9-Decyn-1-ol is an alkyl/ether-based PROTAC linker primarily used in the synthesis of PROTACs. It has been demonstrated to facilitate the conjugation of compounds such as GDC-0068 and Lenalidomide to create highly selective degraders. Additionally, it serves as a valuable click chemistry reagent, utilizing its alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. This product is intended for research use only.
  • Alkyl/ether-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Features an alkyne group for CuAAc reactions
Catalog No. 50-003-40673 Supplier Medchemexpress LLC Supplier No. HY1309855G
9-Decyn-1-ol is an alkyl/ether-based PROTAC linker primarily used in the synthesis of PROTACs. It has been demonstrated to facilitate the conjugation of compounds such as GDC-0068 and Lenalidomide to create highly selective degraders. Additionally, it serves as a valuable click chemistry reagent, utilizing its alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. This product is intended for research use only.
  • Alkyl/ether-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Features an alkyne group for CuAAc reactions