Learn More
New England Biolabs, Inc. 9°N™ DNA Ligase – 2500 units
SDP
Catalog No. 50994899
Encompass
9 N DNA Ligase is a thermostable ligase that catalyzes the formation of a phosphodiester bond between the 5-phosphate and the 3-hydroxyl of two adjacent DNA strands that are hybridized and accurately paired, with no gap, to a complementary DNA strand. 9 N DNA Ligase uses ATP as a cofactor and it is active at elevated temperatures (45 C - 70 C).
Catalog No. 50-994-899 Supplier New England Biolabs, Inc. Supplier No. M0238S
9 N DNA Ligase is a thermostable ligase that catalyzes the formation of a phosphodiester bond between the 5-phosphate and the 3-hydroxyl of two adjacent DNA strands that are hybridized and accurately paired, with no gap, to a complementary DNA strand. 9 N DNA Ligase uses ATP as a cofactor and it is active at elevated temperatures (45 C - 70 C).