Learn More
TARGETMOL CHEMICALS INC 9-dihydro-13-acetylbaccatin II
SDP
Catalog No. 502876655
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 9-dihydro-13-acetylbaccatin III (9-DHAB III)is an apoptosis inducer. It shows cytotoxicity against the MCF7 cell line and drug-resistant cell line MCF7-ADR. Purity 98.96%
Catalog No. 50-287-6655 Supplier TARGETMOL CHEMICALS INC Supplier No. T513225MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 9-dihydro-13-acetylbaccatin III (9-DHAB III)is an apoptosis inducer. It shows cytotoxicity against the MCF7 cell line and drug-resistant cell line MCF7-ADR. Purity 98.96%