Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-Dihydro-13-acetylbaccatin III | 142203-65-4 | 99.6% | C33H42O12 | 25 MG
SDP

Catalog No. 5000358262
Encompass_Preferred

9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate used in the synthesis of paclitaxel analogs. This product appears as a white to off-white solid.

  • Intermediate for paclitaxel analogs
  • High purity of 99.6%
  • Molecular formula C33H42O12
  • Molecular weight 630.68
  • Derived from Taxus chinensis
  • Exhibits antiproliferative activity against P388 cells (IC50 20 μg/mL)

Catalog No. 50-003-58262 Supplier Medchemexpress LLC Supplier No. 25MGHY77434
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate used in the synthesis of paclitaxel analogs. This product appears as a white to off-white solid.

  • Intermediate for paclitaxel analogs
  • High purity of 99.6%
  • Molecular formula C33H42O12
  • Molecular weight 630.68
  • Derived from Taxus chinensis
  • Exhibits antiproliferative activity against P388 cells (IC50 20 μg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.