Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-Dihydro-13-acetylbaccatin III | 142203-65-4 | 99.6% | C33H42O12 | 50 MG
SDP

Catalog No. 5000435016
Encompass_Preferred

9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate used for the preparation of paclitaxel analogs and is intended for research use only. It exhibits antiproliferative activity against mouse P388 cells with an IC50 of 20 μg/mL.

  • Intermediate for the preparation of paclitaxel analogs
  • Intended for research use only
  • Exhibits antiproliferative activity against mouse P388 cells
  • Molecular weight of 630.68
  • Chemical formula is C33H42O12
  • CAS number is 142203-65-4
  • Appears as a white to off-white solid
  • Derived from Taxus chinensis (Pilger) Rehd.

Catalog No. 50-004-35016 Supplier Medchemexpress LLC Supplier No. HY7743450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate used for the preparation of paclitaxel analogs and is intended for research use only. It exhibits antiproliferative activity against mouse P388 cells with an IC50 of 20 μg/mL.

  • Intermediate for the preparation of paclitaxel analogs
  • Intended for research use only
  • Exhibits antiproliferative activity against mouse P388 cells
  • Molecular weight of 630.68
  • Chemical formula is C33H42O12
  • CAS number is 142203-65-4
  • Appears as a white to off-white solid
  • Derived from Taxus chinensis (Pilger) Rehd.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.