Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-Dihydro-13-acetylbaccatin III (9-DHAB III) | 142203-65-4 | 99.6% | C33H42O12 | 1 ML
SDP

Catalog No. 5000435083
Encompass_Preferred

9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate used in the preparation of paclitaxel analogs. This compound exhibits antiproliferative activity against mouse P388 cells, as determined by MTT assay.

  • Intermediate for paclitaxel analog preparation
  • Purity of 99.6%
  • White to off-white solid appearance
  • Classified as terpenoids and diterpenoids
  • Derived from Taxus chinensis plants
  • Demonstrates antiproliferative activity against P388 cells
  • Powder stable for up to 3 years at -20°C or 2 years at 4°C
  • Solution stable for up to 2 years at -80°C or 1 year at -20°C
  • Soluble in DMSO at 20 mg/mL

Catalog No. 50-004-35083 Supplier Medchemexpress LLC Supplier No. HY7743410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate used in the preparation of paclitaxel analogs. This compound exhibits antiproliferative activity against mouse P388 cells, as determined by MTT assay.

  • Intermediate for paclitaxel analog preparation
  • Purity of 99.6%
  • White to off-white solid appearance
  • Classified as terpenoids and diterpenoids
  • Derived from Taxus chinensis plants
  • Demonstrates antiproliferative activity against P388 cells
  • Powder stable for up to 3 years at -20°C or 2 years at 4°C
  • Solution stable for up to 2 years at -80°C or 1 year at -20°C
  • Soluble in DMSO at 20 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.