Learn More
Medchemexpress LLC 9-ethyl-9H-carbazol-3-amine | 132-32-1 | MFCD00004964 | 99.6% | 210.28 g·mol⁻¹ | C14H14N2 | 10 G
SDP
Catalog No. 5000302891
Encompass_Preferred
3-Amino-9-ethylcarbazole (9-ethyl-9H-carbazol-3-amine) is an organic reagent used as a chromogenic substrate in immunohistochemistry and immunoblotting and as a synthetic building block or electronic material in organic electronics research. Supplied at high purity for research, it is suitable for analytical, synthetic, and materials applications.
  • Chromogenic substrate for peroxidase-based staining.
  • High purity suitable for research and synthesis.
  • Used as a building block in organic electronic materials research.
  • Available in laboratory-scale pack sizes for convenient handling.
  • Soluble in common organic solvents for processing and assays.
Catalog No. 50-003-02891 Supplier Medchemexpress LLC Supplier No. HYW01438610G
3-Amino-9-ethylcarbazole (9-ethyl-9H-carbazol-3-amine) is an organic reagent used as a chromogenic substrate in immunohistochemistry and immunoblotting and as a synthetic building block or electronic material in organic electronics research. Supplied at high purity for research, it is suitable for analytical, synthetic, and materials applications.
  • Chromogenic substrate for peroxidase-based staining.
  • High purity suitable for research and synthesis.
  • Used as a building block in organic electronic materials research.
  • Available in laboratory-scale pack sizes for convenient handling.
  • Soluble in common organic solvents for processing and assays.